Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen XVII Antibody (15G9), Novus Biologicals™
SDP

Catalog No. p-200022319 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297682 50 μL
NB297683 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB297682 Supplier Novus Biologicals Supplier No. NBP32664750UL
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Collagen XVII Monoclonal antibody specifically detects Collagen XVII in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen Collagen XVII
Applications Western Blot, ELISA
Classification Monoclonal
Clone 15G9
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias 180 kDa bullous pemphigoid antigen 2, BA16H23.2, BP180alpha 1 type XVII collagen, BPAG2bA16H23.2 (collagen, type XVII, alpha 1 (BP180)), Bullous pemphigoid antigen 2, bullous pemphigoid antigen 2 (180kD), Collagen 17, collagen alpha-1(XVII) chain, collagen XVII, alpha-1 polypeptide, collagen, type XVII, alpha 1, Collagen-17, FLJ60881, KIAA0204, LAD-1
Host Species Rabbit
Immunogen A synthesized peptide derived from Human Collagen XVII [UniProt Q9UMD9]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1308
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.