Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen XXIII alpha 1 Sheep anti-Human, Alexa Fluor 647, Polyclonal, R&D Systems™

Catalog No. IC4165R025
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC4165R025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC4165R025 Supplier R&D Systems Supplier No. IC4165R025
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Collagen XXIII alpha 1 Polyclonal antibody IC4165R025 Specifically detects Collagen XXIII alpha 1 in Human samples. It is validated for Flow Cytometry (Intracellular Staining)

Specifications

Antigen Collagen XXIII alpha 1
Applications Flow Cytometry
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Intracellular Staining by Flow Cytometry 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q86Y22
Gene Alias COL23A1, collagen alpha-1(XXIII) chain, collagen, type XXIII, alpha 1, DKFZp434K0621, procollagen, type XXIII, alpha 1
Gene Symbols COL23A1
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human Collagen XXIII alpha 1 Glu111-Lys540 Accession # Q86Y22
Purification Method Affinity Purified
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91522
Test Specificity Detects human Collagen XXIII alpha 1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human COL25A1 is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.