Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Collagen XXV alpha 1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $427.50
Specifications
Antigen | Collagen XXV alpha 1 |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Description
Collagen XXV alpha 1 Polyclonal specifically detects Collagen XXV alpha 1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Collagen XXV alpha 1 | |
Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
Polyclonal | |
Rabbit | |
Neuroscience | |
PBS with 0.02% Sodium Azide | |
84570 | |
A synthetic peptide mapping to residues 430-443 [DYNGNLHEALQRIT] of the NC3 region of human and mouse CLAC-P. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
80 kDa |
1 mg/mL | |
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
RUO | |
Human | |
Alzheimer disease amyloid-associated protein, AMY, CLAC, CLACP, CLAC-P, Collagen 25, collagen alpha-1(XXV) chain, collagen, type XXV, alpha 1, Collagen-25, collagen-like Alzheimer amyloid plaque component, collagenous Alzheimer amyloid plaque component | |
COL25A1 | |
IgG | |
Affinity Purified | |
This detects full-length CLAC-P and the secreted form of CLAC-P protein. It does not react with full-length or secreted forms of COLXIII protein or the COLXXIII protein. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title