Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen XXV alpha 1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. p-7200701 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018155 0.025 mg
NB300248 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018155 Supplier Novus Biologicals Supplier No. NB300248SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Collagen XXV alpha 1 Polyclonal specifically detects Collagen XXV alpha 1 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen Collagen XXV alpha 1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS with 0.02% Sodium Azide
Gene Alias Alzheimer disease amyloid-associated protein, AMY, CLAC, CLACP, CLAC-P, Collagen 25, collagen alpha-1(XXV) chain, collagen, type XXV, alpha 1, Collagen-25, collagen-like Alzheimer amyloid plaque component, collagenous Alzheimer amyloid plaque component
Gene Symbols COL25A1
Host Species Rabbit
Immunogen A synthetic peptide mapping to residues 155-169 [KGEQGDQGPRMVFPK] of the NC2-2 region of human and mouse CLAC-P.
Molecular Weight of Antigen 80 kDa
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84570
Test Specificity Reacts with residues 155-169 [KGEQGDQGPRMVFPK] of the NC2-2 region of human and mouse CLAC-P. Detects bands of 50, 70 and 100 kDa in SPs of AD brain, but not in diffuse plaques of non-demented aged individuals, diffuse deposits in sub-cortical structures and cerebellum in AD nor in cerebral amyloid angiopathy
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.