Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Collagen XXV alpha 1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | Collagen XXV alpha 1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
Collagen XXV alpha 1 Polyclonal specifically detects Collagen XXV alpha 1 in Human samples. It is validated for Western Blot.Specifications
| Collagen XXV alpha 1 | |
| Polyclonal | |
| Rabbit | |
| Neuroscience | |
| Alzheimer disease amyloid-associated protein, AMY, CLAC, CLACP, CLAC-P, Collagen 25, collagen alpha-1(XXV) chain, collagen, type XXV, alpha 1, Collagen-25, collagen-like Alzheimer amyloid plaque component, collagenous Alzheimer amyloid plaque component | |
| COL25A1 | |
| IgG |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q9BXS0 | |
| 84570 | |
| Synthetic peptides corresponding to COL25A1(collagen, type XXV, alpha 1) The peptide sequence was selected from the middle region of COL25A1. Peptide sequence GERGEAGPPGRGERGEPGAPGPKGKQGESGTRGPKGSKGDRGEKGDSGAQ. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title