Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Collagen XXV alpha 1 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB300249PCP
Description
Collagen XXV alpha 1 Polyclonal antibody specifically detects Collagen XXV alpha 1 in Human, Mouse samples. It is validated for Western BlotSpecifications
Collagen XXV alpha 1 | |
Polyclonal | |
Western Blot | |
Alzheimer disease amyloid-associated protein, AMY, CLAC, CLACP, CLAC-P, Collagen 25, collagen alpha-1(XXV) chain, collagen, type XXV, alpha 1, Collagen-25, collagen-like Alzheimer amyloid plaque component, collagenous Alzheimer amyloid plaque component | |
A synthetic peptide mapping to residues 171-183 [NHGFLSADQQLIK] of the NC2-1 region of human and mouse CLAC-P. | |
0.1 mL | |
Neuroscience | |
84570 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
PerCP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction