Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Collagenase (C. histolyticum)
Shop All Abnova Corporation Products
Click to view available options
Quantity:
100 mg
Description
#REF!
Specifications
Specifications
For Use With (Application) | Enzyme Activity |
Name | Collagenase (C. histolyticum) |
Preparation Method | Native protein purified from Clostridium histolyticum. Prepared cultures grown in medium completely devoid of animal based components and designed for bioprocessing applications where introduction of animal derived pathogens must be prevented. |
Quantity | 100 mg |
Storage Requirements | Store at 4°C on dry atmosphere. For primary cell isolation, culture and tissue dissociation: Reconstituted in balanced salt solutions (such as Hank's, Earle's and others) at concentrations from 0.05% to 0.5% (w/v). For enzymetic assay: Reconstituted in 0.05 M TES buffer with 0.36 mM calcium chloride, pH 7.5 at a concentration of 1.0 mg/ml. |
Regulatory Status | RUO |
Common Name | COLLAGENASE |
Assays | Collagenase Assay; Tissue dissociation. |
Biological Activity | ≥150 units per mg dry weight. One Unit releases one micromole of L-leucine equivalents from collagen in 5 hours at 37°C, pH 7.5. |
Species | Clostridium species |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction