Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagenase, Type I from Clostridium histolyticum

Catalog No. AAJ13820MC
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ13820MC 100 mg
AAJ1382003 1 g
AAJ1382006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ13820MC Supplier Thermo Scientific Supplier No. J13820MC
Add to Cart

Contains mixture of collagenase, clostripain, trypsin, caseinase and other lytic enzymes

  • Contains mixture of collagenase, clostripain, trypsin, caseinase and other lytic enzymes
TRUSTED_SUSTAINABILITY

Specifications

Concentration ≥125 U/mg
Components Contains mixture of collagenase, clostripain, trypsin, caseinase and other lytic enzymes
Content And Storage Store desiccated at 4°C
Source Clostridium histolyticum
Quantity 100 mg
Product Type Collagenase
Form Lyophilized
EINECSNumber 232-582-9
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.