Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vikan™ Collapsible Emergency Blade Shovel

Catalog No. p-7162057 Shop All Remco Products
Click to view available options
Color Code:
Gray
Orange

Collapsible Emergency Blade Shovel has a wide blade that can clean up hazardous materially quickly and easily.

  • Ideal for spill kit applications
  • Constructed of injection-molded ABS plastic
  • Will not damage tires or truck beds
  • It is non-sparking, chemical and corrosion resistant
  • Collapses for easy storage in commercial trucks or vehicles
  • Color-coded

Specifications

Product Type Shovel
Blade Material Polypropylene
Color Code Orange
Color-coded Yes
Handle Material Polypropylene
Head Style Square
Length (English) 36.5 in.
No. per Pack 1 Ea.
Weight (English) 1.77 lb.
Width (English) Head 10.1 in.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.