Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3B Scientific™ Collection of 24 Rocks
SDPSCI_ED

Catalog No. s20725 Shop All 3B Scientific Products
Click to view available options
:
Collection of 24 rocks

The collection contains frequently occurring examples of metamorphic, sedimentary and magmatic rocks as well as important examples of industrial rocks.

Contains:

  • Magmatic rocks, plutonites: foyaite, gabbro, granite, granodiorite, larvikite and monzonite
  • Magmatic rocks, vulcanites: basalt, pumice stone, phonolite, rhyolite
  • Sedimentary rocks: breccia, dolomite, gypsum, limestone, chalk, quartzite and sandstone
  • Metamorphic rocks: amphibolite, eclogite, mica schist, gneiss, marble, phyllite and serpentinite.

Specifications

Product Type Rock collection
Includes 24 rocks
For Use With (Application) Study of Petrography
Subject Earth Science
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.