Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Collibri™ Library Quantification Kit

Catalog No. p-8772007
Encompass_Preferred
Click to view available options
No. of Reactions:
100 Reactions
500 Reactions

Catalog No. A38524100

Supplier: Invitrogen™ A38524100

Only null left
Add to Cart
Add to Cart

Invitrogen™ Collibri™ Library Quantification Kit includes a ready-to-use master mix optimized for Illumina NGS library quantification and a library dilution buffer

  • Includes Platinum II Taq Hot Start DNA Polymerase and enables convenient and accurate quantification of NGS libraries
  • Contains all components needed for the amplification reaction, including primers and passive reference dye, and does not require any preparation before use
  • Due to the universal concentration of the passive reference dye in the master mix, the kit can be used with any qPCR instrument without adjustments
  • Accurate quantification of libraries made from various sample types, including degraded samples
  • Ease of use with all components ready to use -- no reagent preparation required
  • Visual cues - specially formulated dyes included to track pipetting steps
  • Platinum hot-start technology - superior specificity, sensitivity, and room-temperature reaction setup Unmatched convenience
  • Contains:
    • Platinum II Taq Hot Start DNA Polymerase
    • Optimized buffer
    • dNTPs
    • Primer mix targeting adapter sequences
    • SYBR green dye
    • Passive reference dye
    • Blue dye to track pipetting steps
  • Unique formulation provides ease of use at every step of the workflow:
    • No reagent preparation
    • Tracking of pipetting steps
    • One cycling protocol for libraries of various lengths and GC/AT-content levels
    • Universal passive reference dye concentration - a single master mix for all qPCR instruments
  • Also includes a ready-to-use library dilution buffer that contains stabilizing agents to enable library integrity after dilution. The buffer contains yellow dye to track pipetting steps. When combined, the blue master mix and yellow diluted libraries become a green solution. This feature eases tracking of the addition of the diluted libraries or DNA standards to the master mix in the qPCR plate wells, helping reduce pipetting errors.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -20°C.
Detection Method SYBR
Format Kit
Sample Type DNA, RNA
For Use With (Equipment) 7500 Fast System, 7900HT Fast System, QuantStudio™, ViiA™ 7 System, StepOne™ System, StepOnePlus™ System
GC-Rich PCR Performance Low
For Use With (Application) Illumina NGS Library Quantification
No. of Reactions 100 Reactions
PCR Method qPCR
Polymerase Platinum™ II Taq Hot Start DNA Polymerase
Product Line Collibri
Product Type Library Quantification Kit
Quantity 100 reactions
Shipping Condition Dry Ice
Test Time Standard
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.