Learn More
Collision in 2-D Apparatus
Click to view available options
:
Collision In 2D apparatus
Description
- Includes heavy-gauge aluminum launching platform with base, two 1.2cm steel balls, one 2.5cm wooden ball and one 1.2cm glass ball
- Movable target arm offers flexibility to conduct broad range of collision experiments
- Attaches easily to a stand or table edge with a C-clamp (S49174)
- Instructions included
Specifications
Specifications
Product Type | Collision In 2D Apparatus |
For Use With (Application) | Used for studying the conservation of momentum and kinetic energy in both elastic and inelastic collisions in two dimensions |
Includes | Heavy gauge aluminum platform, two 1.2cm steel ball, one 1.2cm glass ball, one 2.5cm wood balls and instructions |