Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Collybistin/ARHGEF9 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | Collybistin/ARHGEF9 |
---|---|
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Collybistin/ARHGEF9 Polyclonal specifically detects Collybistin/ARHGEF9 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Collybistin/ARHGEF9 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
Cdc42 guanine nucleotide exchange factor (GEF) 9, COLLYBISTIN, EIEE8, hPEM-2 collybistin, KIAA0424HPEM-2, PEM2, PEM-2, PEM-2 homolog, Rac/Cdc42 guanine nucleotide exchange factor 9, rho guanine nucleotide exchange factor 9 | |
ARHGEF9 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Polyclonal | |
Rabbit | |
Human | |
O43307 | |
23229 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:DEEGWFPASFVRLWVNQEDEVEEGPSDVQNGHLDPNSDCLCLGRPLQNRDQMRANVINEIMSTERHYIKHLKDIC | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title