Learn More
R&D Systems™ Colon Matched Tumor and Normal Tissue Lysate
SDP
Catalog No. NB002092 Shop All Novus Products
Change view
Click to view available options
Quantity:
2 Vials
Catalog No. Quantity
NB002092 2 Vials
1 options
Catalog No. NB002092 Supplier R&D Systems™ Supplier No. NBP2470722VIAL
Add to Cart
Add to Cart
A Novus product, part of R&D Systems. Two matched 1 mg/mL vials (100 μg/vial) of human colon tissue lysate, one containing normal tissue and the other tumor tissue, for use in research. Lysates are guaranteed for 6 months from date of receipt.
The Colon Matched Tumor and Normal Tissue Lysate is derived from human.
Specifications
Quantity 2 Vials
Content And Storage Store at -80C. Avoid freeze-thaw cycles.
Product Line Novus