Learn More
American Educational Products Color-Coded Spring Scales
Click to view available options
Color:
Multiple
White
Description
Series of six color-coded tubular spring scales can be used for both force and mass measurements.
- Calibrated in both Newtons and grams with a beveled top for use on an inclined plane
- Incorporate a unique zero adjustment with a clear plastic tube, allowing students to see spring action
Order Info
Available separately or as a set of all six colors
Specifications
Specifications
Color | White |
Capacity | 3 kg, 30 N |
Type | Spring Scale |