Learn More
Hand2Mind Color Mix Sensory Tubes

Description
- From the company that specializes in learning manipulatives for kids, try our sensory fidget tubes that help your child deal with social emotional learning.
- Help your students manage their emotions, focus their attention and practice quiet, calm engagement with our 3 ColorMix Sensory Fidget Tubes.
- These securely sealed, easy-to-grip bottles offer fascinating movement patterns and provide a quiet option for students who need help settling down.
- For over 50 years, hand2mind has encouraged hands-on learning and discovery, driving deeper understanding and helping children unlock their full potential. Hand2mind is a pioneer in social emotional learning products that help kids build mindfulness as well as how to deal with their own emotions.
- From kindergarten to middle school, hand2mind has you covered with products designed to enhance learning in the classroom and the world around us.
- Through hands-on exploration and discovery we strive to find new ways to encourage students in math practice, literacy activities, STEM education, Social Emotional Learning and more!
Specifications
Specifications
| Grade Level | Elementary, Intermediate - Middle School, High School |
| Grade | K to 12 |
| Class Size | 1 Student |
| Product Type | Sensory Tubes |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.