Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EP Reference Solution Y5, Yellow Reference Solution, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
120 mL
Specifications
Specifications
Chemical Name or Material | Color standard Y5 |
Packaging | Amber Glass Bottle |
CAS | 7732-18-5, 7647-01-0, 7758-99-8, 10025-77-1, 7791-13-1, 7647-01-0, 7732-18-5 |
Color | Yellow |
pH | Acidic |
Melting Point | 0.0°C |
Boiling Point | 100°C |
For Use With (Equipment) | Water Analyzer |
Synonym | Yellow standard solution |
Solubility | Miscible with water |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction