Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Colorblindness Testbook



Click to view available options
Format:
Book
Description
- Set of 14 test cards compiled by Dr. S. Ishihara Explanation booklet included
- Designed to help determine the type and severity of color blindness in a patient
Specifications
Specifications
Author | Dr. S. Ishihara |
Format | Book |
Product Type | Colorblindness Testbook |
For Use With (Application) | For color deficiency |
Subject | Biology |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction