Learn More
Fisherbrand™ Colormark™ Plus Adhesion Slides
Shop All Fisherbrand Products
Click to view available options
Description:
Colormark Plus
Description
Fisherbrand™ Colormark Slides are brightly coated with contrasting background color. Background color makes etched patient information more visible.
Designed for use with automated slide labeling machines. Offer excellent etched labelling visibility.
- Self-contained packages ready for loading into your slide labeling system
- Simply remove the cover from the slide package and drop the sleeve into the loading bin of the labeling system
- Available with Plus formula to provide the utmost adhesion
Specifications
Specifications
Type | Adhesion Slide |
Material | Glass |
Frosted | Yes |
Precleaned | Yes |
Color | Plain |
Description | Colormark Plus |