Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ COLQ Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA530941
Description
Recommended positive controls: HepG2. Predicted reactivity: Mouse (87%), Rat (82%), Bovine (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
COLQ (collagen-like tail subunit (single strand of homotrimer) of asymmetric acetylcholinesterase), also known as EAD, is a 455 amino acid protein that localizes to the end plate of skeletal muscle. COLQ anchors the catalytic subunits of asymmetric AChE (acetylcholinesterase) to the basal lamina at the neuromuscular junctions of vertebrates. Mutations of COLQ lead to congenital myasthenic syndromes which are rare autosomal recessive diseases characterized by general weakness increased by exertion, ophthalmoplegia and refractoriness to anticholinesterase drugs. Eight isoforms exist due to alternative splicing events.
Specifications
| COLQ | |
| Polyclonal | |
| Unconjugated | |
| COLQ | |
| A130034K24Rik; acetylcholinesterase collagenic tail peptide; Acetylcholinesterase-associated collagen; AChE Q subunit; CMS5; collagen like tail subunit of asymmetric acetylcholinesterase; collagenic tail of endplate acetylcholinesterase; collagen-like tail subunit (single strand of homotrimer) of asymmetric acetylcholinesterase; collagen-like tail subunit of asymmetric acetylcholinesterase; Colq; EAD; single strand of homotrimeric collagen-like tail subunit of asymmetric acetylcholinesterase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8292 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9Y215 | |
| COLQ | |
| Recombinant protein encompassing a sequence within the center region of human COLQ. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction