Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COLQ Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575768
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575768 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575768 Supplier Invitrogen™ Supplier No. PA575768
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

COLQ (collagen-like tail subunit (single strand of homotrimer) of asymmetric acetylcholinesterase), also known as EAD, is a 455 amino acid protein that localizes to the end plate of skeletal muscle. COLQ anchors the catalytic subunits of asymmetric AChE (acetylcholinesterase) to the basal lamina at the neuromuscular junctions of vertebrates. Mutations of COLQ lead to congenital myasthenic syndromes which are rare autosomal recessive diseases characterized by general weakness increased by exertion, ophthalmoplegia and refractoriness to anticholinesterase drugs. Eight isoforms exist due to alternative splicing events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COLQ
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene COLQ
Gene Accession No. O35167, O35348, Q9Y215
Gene Alias A130034K24Rik; acetylcholinesterase collagenic tail peptide; Acetylcholinesterase-associated collagen; AChE Q subunit; CMS5; collagen like tail subunit of asymmetric acetylcholinesterase; collagenic tail of endplate acetylcholinesterase; collagen-like tail subunit (single strand of homotrimer) of asymmetric acetylcholinesterase; collagen-like tail subunit of asymmetric acetylcholinesterase; Colq; EAD; single strand of homotrimeric collagen-like tail subunit of asymmetric acetylcholinesterase
Gene Symbols COLQ
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues in Human COLQ.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29755, 382864, 8292
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.