Description
- Jacket permits column to be used as condenser
- With top outer and lower inner standard taper joints
Specifications
Specifications
| Height (Metric) | 280mm |
| Type | Distillation Condenser |
| Standard Taper Joint | 14/20 |