Learn More
Thermo Scientific™ Column Extenders for Pierce™ Centrifuge Columns PROMO
Description
Pierce Centrifuge Columns are convenient tools for chromatography experiments involving 40μL to 10 mL of beaded agarose or other affinity matrix. The columns are made from low protein binding polypropylene for compatibility with antibody and other protein purification, and they fit into standard centrifuge tubes for use in any centrifuge. Simply add the affinity resin to one of the-polypropylene columns, remove the twist-off bottom and allow the beads to settle to form the resin bed. Process sample by gravity-flow, then use a centrifuge to efficiently wash and elute purification products from the column.
Pierce Centrifuge Columns can be used in either spin or gravity-flow format. The spin format reduces sample processing time and makes column washing and elution more efficient. For gravity-flow procedures and when slow processing time is desired for binding steps with dilute samples, Column Extenders can be used to increase the column reservoir capacity. Use any combination of processing techniques to get the best possible chromatography results!
Highlights:
- Available in four sizes to accommodate 40μL to 10 mL resin volumes
- Perform experiments using any combination of spin or gravity-flow processing
- Columns made of polypropylene with polyethylene filter discs, ensuring minimal protein-binding properties
- Columns fit into standard centrifuge tubes for sample collection and centrifugation in standard benchtop centrifuge rotors
0.8mL Centrifuge Columns (Part No. 89868, 89869)
- Total volume: 800μL
- Resin volume: 40 to 400μL
- Filter type: polyethylene filter, ∽30μm (20 to 50) pore size
- Receiver tube: fits standard microcentrifuge tubes
- Cap type: O-ring screw top caps; twist-off bottoms
2 mL Centrifuge Columns (Part No. 89896)
- Total volume: 5 mL (2 mL resin bed, 3 mL reservoir)
- Resin volume: 2 mL
- Filter type: polyethylene filter, ∽30μm (20 to 50) pore size
- Receiver tube: fits standard 15 mL conical centrifuge tubes
- Cap type: screw top caps; twist-off bottoms and press-on caps to reseal
5 mL Centrifuge Columns (Part No. 89897)
- Total volume: 8mL (5 mL resin bed, 3 mL reservoir)
- Resin volume: 5 mL Filter type: polyethylene filter, ∽30μm (20 to 50) pore size
- Receiver tube: fits standard 15 mL conical centrifuge tubes
- Cap type: screw top caps; twist-off bottoms and press-on caps to reseal
10 mL Centrifuge Columns (Part No. 89898)
- Total volume: 22 mL (10 mL resin bed, 12 mL reservoir)
- Resin volume: 10 mL
- Filter type: polyethylene filter, ∽30μm (20 to 50) pore size
- Receiver tube: fits standard 50 mL conical centrifuge tubes
- Caps: screw top caps; twist-off bottoms and press-on caps to reseal
Column Extenders (Part No. 69707)
- Increase column reservoir volume by 35 to 40 mL
- Designed especially for use in gravity-flow procedures
- Fit Part No. 89896, 89897, 89898 only
Specifications
Specifications
| Capacity (Metric) | 35 mL |
| Content And Storage | Store at room temperature. |
| Product Line | Pierce™ |
| Quantity | 10 Extenders |
| Type | Column Accessory |
| Format | 15 mL Conical, Accessory |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.