Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Com-Pac International™ SAF-T-ZIP™ Specimen Bags
Click to view available options
Color:
Green
Red
Description
- Color-coded zippers for easy sorting
- Convenient document pouch separates documents from the specimen
- Printed with the OSHA-required biohazard symbol
- Measure 6 × 9 in. (15.2 × 22.9cm)
Specifications
Specifications
Closure Type | Zip Seal |
Color | Red |
Dimensions (L x W) | 6 x 9 in. (15.2 x 22.9cm) |
Length (English) | 6 in. |
Length (Metric) | 15.2 cm |
Width (English) | 9 in. |
Width (Metric) | 22.9 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction