Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Comanic Acid 98.0+%, TCI America™
SDP

Catalog No. C110425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C110425G 25 g
C11041G 1 g
C11045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C110425G Supplier TCI America Supplier No. C110425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 499-05-8
Molecular Formula C6H4O4
Molecular Weight (g/mol) 140.094
MDL Number MFCD00059958
InChI Key UFMDRPPBUAPWBK-UHFFFAOYSA-N
Synonym 4-Oxo-4H-pyran-2-carboxylic Acid, gamma-Pyrone-2-carboxylic Acid
PubChem CID 12305536
IUPAC Name 4-oxopyran-2-carboxylic acid
SMILES C1=COC(=CC1=O)C(=O)O

Specifications

Quantity 25 g
Formula Weight 140.09
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Comanic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.