Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CombiMethanol Solvent, MilliporeSigma™

Catalog No. p-7162448 Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 L
2.5 L

CombiMethanol Solvent for volumetric Karl Fischer titration with one component reagents max. 0.01% H2O Aquastar™.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point −98°C
Boiling Point 64.5°C (1013 hPa)
Density 0.792 g/cm3 (at 20°C)
CAS 67-56-1
Quantity 1 L
Flash Point 9.7°C
Molecular Formula CH4O
Packaging Glass Bottle
Synonym MeOH, Hydroxymethane, Methyl alcohol, Carbinol, Hydroxymethane, Methyl alcohol, Carbinol
Molecular Weight (g/mol) 32.04
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.