Learn More
Dentec Safety™ Combination Gas Cartridge
SDP
Catalog No. p-7162495 Shop All Dentec Safety Specialists Inc. Products
Click to view available options
Filter Type:
Combination Acid Gases and P100
Combination Acid Gases/Organic Vapor and P100
Combination Ammonia/Methylamine and P100
Combination Multi-Vapor Gas and P100
Combination Organic Vapor and P100
Quantity:
4/Box
60/Box
Catalog No. 19-222-519
Supplier: Dentec Safety™ 158T21LP0
Only null left
Add to Cart
Add to Cart
Dentec Safety™ Combination Gase Cartridge features unique space plenums that allow users to access the entire filter material reducing breathing resistance, providing longer cartrdige life, reducing costs and improving productivity
  • Offset connector allows the user to rotate the cartridge into 3 different positions improving visibility
  • Saf-Twist design quickly attaches the cartridge to any Dentec Safety half-mask or full face air-purifying respirator Facepiece.
Specifications
Filter Type Combination Acid Gases and P100
Material Plastic
Product Type Respirator Accessories
For Use With (Equipment) Masks
Includes Cartridges and filters
Quantity 4/Box
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.