Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ Combitips™ Advanced Tips for Multipette™ Multi-dispensers - Forensic DNA Grade
GREENER_CHOICE

Catalog No. p-7112266
Click to view available options
Volume (Metric):
1 mL
2.5 mL
5 mL

Redesigned and optimized to meet needs of modern laboratory. Eppendorf™ Combitips advanced™ Pipette Tips - Forensic DNA Grade are quick at dispensing of identical volumes.

  • Ideal system components for all Eppendorf™ multipettes
  • Offers range of dispensing volumes with increments as low as 100 nanoliters
  • Elongated combitips in volumes 2.5 mL and 5 mL make it possible to reach bottom of laboratory vessels
  • Individual color coding facilitates quick identification of desired combitips
  • Unique funnel geometry ensures comfortable handling while preventing damage to gloves

Applications

  • Positive displacement principle
  • High-precision dispensing regardless of physical properties of liquid (eg: viscosity, volatility, density, temperature)
  • Prevents aerosol contamination with hermetically sealed piston for secure dispensing
  • Provides protection from radioactive and toxic substances
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1 mL
Sterility Non-sterile
Color Colorless
Autoclavable Not Autoclavable
Clean Claims Forensic DNA Grade (non-sterile)
Format Individually Wrapped
Tip Style Extended Length, Positive Displacement
Packaging 100 Pk., Individually Blister-wrapped
Product Type Pipette Tips
Color Code Yellow
Disposable Single-use
For Use With (Equipment) Eppendorf Multipette
Graduated Yes
Product Line CombiTips Advanced
Material Polypropylene
Fit for Purpose Forensic Ready
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.