Learn More
Description
Includes
Lysis Solution, 2 x 500mL; Comet LMAgarose, 15mL; Trevigen CometSlide, 25/kit; pH 10 200mM EDTA, 12.5mL; Kit with Silver Stain (Mfr. No. 4251-050-K) also includes four staining reagents and a fixation additive.
The principle of the assay is based upon the ability of denatured, cleaved DNA fragments to migrate out of the nucleoid under the influence of an electric field, whereas undamaged DNA migrates slower and remains within the confines of the nucleoid when a
Choose the Neutral CometAssay to detect double-stranded breaks or the Alkaline CometAssay for a more sensitive protocol used to detect smaller amounts of damage including single and double-stranded breaks
Cells are immobilized in a bed of low melting poi
Specifications
Specifications
| Quantity | 50 Tests |
| Product Type | Single Cell Gel Electrophoresis Assay |
| Content And Storage | Lysis Solution, Comet LMAgarose, CometSlide, pH 10 200mM EDTA, four staining reagents and fixation additive, Silver Stain. The product requires storage at room temperature, 2 - 8° C, and -20° C. Consult the product insert for specific storage temperatures. Do not use past expiration date. |
| Label or Dye | Silver Stain |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
