Learn More
R&D Systems™ CometChip™ Electrophoresis Starter Kit
Catalog No. p-7162867
Click to view available options
Product Type:
ES Starter Kit+NA Power Supply
This item is not returnable. View return policy
Caution! Electrical Hazard! This equipment is designed for use with a DC power supply providing up to 250 VDC. Careless handling can result in electrical shock. To avoid any risk of injury, the instrument should be operated in accordance with the instruction provided.
This item is not returnable. View return policy
Caution! Electrical Hazard! This equipment is designed for use with a DC power supply providing up to 250 VDC. Careless handling can result in electrical shock. To avoid any risk of injury, the instrument should be operated in accordance with the instruction provided.
High-throughput platform to treat and measure DNA damage on a single slide
  • Includes:
    • CometChip™ Starter Kit
    • CometAssay™ Electrophoresis System II
Specifications
Content And Storage The product requires storage at both 2 - 8°C and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date.
Includes CometAssay™ Electrophoresis System II, 96-Well CometChip™ System, CometChip™ 30 micron, 100 mL Lysis Solution, & 15 mL Comet LMAgarose
Product Type ES Starter Kit+NA Power Supply
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.