Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ CometSlide™
Click to view available options
Quantity:
2 Slides
25 Slides
Description
- A component of the CometAssay™ Kit , which is used for the rapid analysis of DNA fragmentation associated with DNA damage
- The CometAssay™, or single cell gel electrophoresis assay, permits the quantification of DNA damage in individual cells based on the migration of DNA structures in an electric field
- Evaluation of the DNA Comet tail shape and migration pattern allows for assessment of DNA damage, expressed as tail length, percent DNA in the tail and tail moment
- CometSlide™ provides a convenient, simple, and reliable format for sample preparation, processing, analysis, and storage for the CometAssay™
Specifications
Specifications
Content And Storage | Store the unopened product at room temperature. Do not use past expiration date. |
Description | Sample-ready pre-treated glass slides |
For Use With (Equipment) | CometAssay™ Kit |
For Use With (Application) | Sample preparation, processing, analysis, and storage for the CometAssay™ |
Quantity | 2 Slides |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction