Learn More
R&D Systems™ CometSlide
Description
Convenient, simple and reliable sample preparation, processing, analysis and storage format for the CometAssay kits
Features a writing surface, specially treated glass surface and hydrophobic barrier for sample
Specifications
Specifications
| Quantity | 100 Slides |