Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dentec Safety™ Comfort-Air™ Series 100 Half-Mask Respirators
SDP

Catalog No. p-7162508 Shop All Dentec Safety Specialists Inc. Products
Click to view available options
Size:
Small
Medium
Large

Pure silicone half-mask respirators are comfortable and easy to clean.

  • Extended field of vision
  • Large inhalation valves
  • Exhalation valve is strategically positioned under the nostrils and in front of the mouth to ensure hot, exhaled air exits the mask immediately
  • Extra Large face seal surface offers comfort
  • Textured finish on the sealing area reduces the risk of the mask slipping on the face
  • Enhanced harness assembly features:
    • Metal tabs at the end to prevent nylon strap from fraying and allowing for adjustment when wearing gloves
    • Wide nylon strap maintains elasticity longer.
    • 4-point swiveling adjustments
    • Cradle suspension fits comfortably under suspension systems for hard hats, grinding and welding helmets
  • 3-position adjustment allows users to adjust cartridge to three different positions

Specifications

Certifications (NIOSH) NIOSH 42 CFR Part 84
Color Blue
Material Silicone
Product Type Half-Mask Respirator
Size Small
With Exhalation Valve Yes
Facepiece Material Silicone
For Use With (Equipment) Dentec™ Filters and Cartridges
Material Headstrap Nylon
Model 100-S-00
Length (Metric) 8.89 cm
Width (Metric) 17.78 cm
Height (Metric) 12.7 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.