Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dentec Safety™ Comfort-Ease AD4N95 Flat-Fold N95 Respirators

Flat-fold respirators feature mask flexibility that moves with the jaw, allowing it to maintain a proper, secure seal.
Specifications
Aerosol Type | N - Not Resistant to Oil |
---|---|
Certifications (NIOSH) | NIOSH 42 CFR 84, N95 |
Design Type | Cup/Molded |
Disposable | Single-use |
Filter Type | N95 |
Catalog Number | Mfr. No. | With Exhalation Valve | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | With Exhalation Valve | Price | Quantity & Availability | |||||
19-222-403
![]() |
Dentec Safety™
AD4N95B240 |
No | N/A | N/A | N/A | ||||
19-222-404
![]() |
Dentec Safety™
AD4N95VB120 |
Yes | N/A | N/A | N/A | ||||
Description
- Flat fold design for ideal comfort and fit and allows the user to store the product in a pocket
- Extended field of vision
- Soft material and soft foam seal at nose bridge improves user comfort
- Pass-through straps:
- Allow for quick and simple adjustment
- Allows user to remove crown strap and conveniently hang around the neck
- Ideal for users required to move from a hazardous area to non hazardous area
- Keeps the mask with the user
- Reduces consumption costs
- Adjustable aluminum bar at nose bridge allows user to fit the mask quickly and easily.
- Aluminum bar is sewn into material for dielectric applications
Specifications
N - Not Resistant to Oil | |
Cup/Molded | |
N95 | |
Standard | |
Yes | |
Individually Packaged | |
13.97 cm | |
No |
NIOSH 42 CFR 84, N95 | |
Single-use | |
General Purpose Particulate Respirator | |
95% | |
Adjustable Nose Clip | |
Non-sterile | |
Single Continuous Strap |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title