Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COMMD1 Antibody (OTI2E2), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272418F
Description
COMMD1 Monoclonal specifically detects COMMD1 in Human, Mouse, Rat, Monkey samples. It is validated for Flow Cytometry, Immunohistochemistry.Specifications
COMMD1 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
COMMD1 | |
Full length recombinant protein of human COMMD1 (NP_689729) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Monkey | |
IgG2a |
Flow Cytometry, Immunohistochemistry | |
OTI2E2 | |
Flow Cytometry, Immunohistochemistry | |
C2orf5, COMM domain-containing protein 1, copper metabolism (Murr1) domain containing 1, copper metabolism gene MURR1, MGC27155, MURR1chromosome 2 open reading frame 5 (MURR1), Protein Murr1 | |
Mouse | |
Affinity Purified | |
Signal Transduction | |
150684.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction