Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COMMD1 Antibody (OTI2E2), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272418MFV610
Description
COMMD1 Monoclonal antibody specifically detects COMMD1 in Human, Mouse, Rat, Monkey samples. It is validated for Flow CytometrySpecifications
COMMD1 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Monkey | |
Purified |
Flow Cytometry | |
OTI2E2 | |
Flow Cytometry | |
C2orf5, COMM domain-containing protein 1, copper metabolism (Murr1) domain containing 1, copper metabolism gene MURR1, MGC27155, MURR1chromosome 2 open reading frame 5 (MURR1), Protein Murr1 | |
Full length human recombinant protein of human COMMD1(NP_689729) produced in HEK293T cell. | |
0.1 mL | |
Signal Transduction | |
150684 | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction