Learn More
Invitrogen™ COMMD1 Monoclonal Antibody (2B12)

Description
Peptide Sequence: MAAGELEGGK PLSGLLNALA QDTFHGYPGI TEELLRSQLY PEVPPEEFRP FLAKMRGILK SIASADMDFN QLEAFLTAQT KKQGGITSDQ AAVISKFWKS HKTKIRESLM NQSRWNSGLR GLSWRVDGKS QSRHSAQIHT PVAIIELELG KYGQESEFLC LEFDEVKVNQ ILKTLSEVEE SISTLISQPN.
Specifications
Specifications
| Antigen | COMMD1 |
| Applications | ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 2B12 |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | COMMD1 |
| Gene Accession No. | Q8N668 |
| Gene Alias | AI256843; C2orf5; COMM domain containing 1; COMM domain-containing protein 1; Commd1; copper metabolism (Murr1) domain containing 1; copper metabolism domain containing 1; copper metabolism gene MURR1; Murr1; Protein Murr1; U2/Mu; U2af1-rs1 region 1 |
| Gene Symbols | COMMD1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.