Learn More
Invitrogen™ COMMD10 Polyclonal Antibody

Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: SLEKQDLHLV LETISFILEQ AVYHNVKPAA LQQQLENIHL RQDKAEAFVN TWSSMGQ Highest antigen sequence indentity to the following orthologs: Rat - 84%, Mouse - 86%.
Specifications
Specifications
| Antigen | COMMD10 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | COMMD10 |
| Gene Accession No. | Q9Y6G5 |
| Gene Alias | 2310003A05Rik; AU019438; COMM domain containing 10; COMM domain-containing protein 10; COMMD10; Down-regulated in W/WV mouse stomach 2; DRWMS2; HSPC305; mDRWMS2; PTD002 |
| Gene Symbols | COMMD10 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.