Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COMMD2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP191802
Description
COMMD2 Polyclonal specifically detects COMMD2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
COMMD2 | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
COMM domain containing 2, COMM domain-containing protein 2, HSPC042, MGC57611 | |
Rabbit | |
Affinity Purified | |
RUO | |
51122 | |
Human, Mouse, Rat | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
COMMD2 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:FVLGFSEELNKLLLQLYLDNRKEIRTLLSELAPSLPSYHNLEWRLDVQLASRSLRQQIKPAVTIKLHLNQNGDHNTKVLQTDPATLLHLVQQLEQALE | |
0.1 mL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction