Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ COMMD2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570248
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 92%.
COMMD2 may modulate activity of cullin-RING E3 ubiquitin ligase (CRL) complexes. may down-regulate activation of NF-kappa-B.
Specifications
COMMD2 | |
Polyclonal | |
Unconjugated | |
COMMD2 | |
1190017G07Rik; AI047739; COMM domain containing 2; COMM domain-containing protein 2; COMMD2; D3Ertd176e; HSPC042; My004; wu:fb12e01; zgc:92665 | |
Rabbit | |
Affinity chromatography | |
RUO | |
688478 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
0 | |
COMMD2 | |
synthetic peptide corresponding to a region of rat COMMD2. | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction