Learn More
Invitrogen™ COMMD3 Monoclonal Antibody (2E2)

Description
Peptide Sequence: MELSESVQKG FQMLADPRSF DSNAFTLLLR AAFQSLLDAQ ADEAVLDHPD LKHIDPVVLK HCHAAAATYI LEAGKHRADK STLSTYLEDC KFDRERIELF CTEYQNNKNS LEILLGSIGR SLPHITDVSW RLEYQIKTNQ LHRMYRPAYL VTLSVQNTDS PSYPEISFSC SMEQLQDLVG KLKDASKSLE RATQS.
Specifications
Specifications
| Antigen | COMMD3 |
| Applications | ELISA, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 2E2 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | COMMD3 |
| Gene Accession No. | Q9UBI1 |
| Gene Alias | AW550818; Bmi1 upstream; bmi-1 upstream gene protein; Bup; Bup protein; C10orf8; COMM domain containing 3; COMM domain-containing protein 3; COMMD3; D2Ertd542e; Protein Bup; Protein PIL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.