Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COMMD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557730
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557730 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557730 Supplier Invitrogen™ Supplier No. PA557730
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MELSESVQKG FQMLADPRSF DSNAFTLLLR AAFQSLLDAQ ADEAVLDHPD LKHIDPVVLK HCHAAAATYI LEAGKHRADK STLSTYLEDC KFDRE Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 89%.

COMMD3 may modulate activity of cullin-RING E3 ubiquitin ligase (CRL) complexes. may down-regulate activation of NF-kappa-B. Modulates Na(+) transport in epithelial cells by regulation of apical cell surface expression of amiloride-sensitive sodium channel (ENaC) subunits.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COMMD3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene COMMD3
Gene Accession No. Q9UBI1
Gene Alias AW550818; Bmi1 upstream; bmi-1 upstream gene protein; Bup; Bup protein; C10orf8; COMM domain containing 3; COMM domain-containing protein 3; COMMD3; D2Ertd542e; Protein Bup; Protein PIL
Gene Symbols COMMD3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human COMMD3. Recombinant protein control fragment (Product #RP-97101).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23412
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.