Learn More
Invitrogen™ COMMD5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595782
Description
Immunogen sequence: MSAVGAATPY LHHPGDSHSG RVSFLGAQLP PEVAAMARLL GDLDRSTFRK LLKFVVSSLQ GEDCREAVQR LGVSANLPEE QLGALLAGMH TLLQQALRLP PTSLKPDTFR DQLQELCIPQ DLVGDLASVV FGSQRPLLDS VAQQQGAWLP HVADFRWRVD VAISTSALAR SLQPSVLMQL KLSDGSAYRF EVPTAKFQEL RYSVALVLKE MADLEKRCER RLQD; Positive Samples: SGC996, 293T, LO2, HeLa, A-431, Mouse kidney, Rat liver; Cellular Location: Cytoplasm, Nucleus.
COMMD5 is a protein coding gene. May modulate activity of cullin-RING E3 ubiquitin ligase (CRL) complexes. Negatively regulates cell proliferation. Negatively regulates cell cycle G2/M phase transition probably by transactivating p21/CDKN1A through the p53/TP53-independent signaling pathway. Involved in kidney proximal tubule morphogenesis. Down-regulates activation of NF-kappa-B.
Specifications
COMMD5 | |
Polyclonal | |
Unconjugated | |
COMMD5 | |
2310065H03Rik; AI854466; COMM domain containing 5; COMM domain-containing protein 5; COMMD5; D15Ertd81e; FLJ13008; HCARG; HT002; hypertension-related calcium regulated; hypertension-related calcium-regulated gene protein; hypertension-related, calcium regulated | |
Rabbit | |
Affinity chromatography | |
RUO | |
245974, 28991, 66398 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1.48 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
Q8R395, Q9ERR2, Q9GZQ3 | |
COMMD5 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-224 of human COMMD5 (NP_0547852). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.