Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COMMD5 Polyclonal Antibody
COMMD5 Polyclonal Antibody
Supplier: Thermo Scientific PA5100232
Description
COMMD5 Polyclonal Antibody for Western Blot, ICC/IF

Specifications
COMMD5 | |
Polyclonal | |
Unconjugated | |
COMMD5 | |
2310065H03Rik; AI854466; COMM domain containing 5; COMM domain-containing protein 5; COMMD5; D15Ertd81e; FLJ13008; HCARG; HT002; hypertension-related calcium regulated; hypertension-related calcium-regulated gene protein; hypertension-related, calcium regulated | |
Rabbit | |
Affinity Chromatography | |
RUO | |
245974, 28991, 66398 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8R395, Q9ERR2, Q9GZQ3 | |
COMMD5 | |
A synthesized peptide derived from human COMMD5(Accession Q9GZQ3), corresponding to amino acid residues M1-L43. | |
100 μL | |
Primary | |
Human, Mouse, Rat, Monkey | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction