Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COMMD5 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP310847100UL
Description
COMMD5 Polyclonal specifically detects COMMD5 in Mouse samples. It is validated for Western Blot.Specifications
COMMD5 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
COMM domain containing 5, COMM domain-containing protein 5, FLJ13008, HCARG, HT002, Hypertension-related calcium-regulated gene protein, MGC72046 | |
The immunogen is a synthetic peptide directed towards the middle region of mouse COMMD5 (NP_079812.1). Peptide sequence GIPQDMIGDLASLAFGSQRPLLDSVAQQQGSSLPRVSNFRWRVDVAISTS | |
100 μg | |
Primary | |
Mouse | |
Purified |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
28991 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction