Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COMMD7 Antibody (49B5), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259423AF532
This item is not returnable.
View return policy
Description
COMMD7 Monoclonal antibody specifically detects COMMD7 in Human samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
COMMD7 | |
Monoclonal | |
Alexa Fluor 532 | |
C20orf92, chromosome 20 open reading frame 92, COMM domain containing 7, COMM domain-containing protein 7, dJ1085F17.3, FLJ14987, MGC33315 | |
Recombinant human COMMD7(1-200aa)purified from E. coli | |
0.1 mL | |
Cancer, Cell Cycle and Replication | |
149951 | |
Store at 4°C in the dark. | |
IgG3 κ |
Western Blot, ELISA, Immunofluorescence | |
49B5 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction