Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COMMD7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565840
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565840 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565840 Supplier Invitrogen™ Supplier No. PA565840
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LHCTEDPVPEA VGGDMQQLNQ LGAQQFSALT EVLFHFLTEP KEVERFLAQL SEFATTNQIS LGSLRSIVKS Highest antigen sequence identity to the following orthologs - mouse 90%, rat 93%.

May modulate activity of cullin-RING E3 ubiquitin ligase (CRL) complexes (PubMed:21778237). Associates with the NF-kappa-B complex and suppresses its transcriptional activity (PubMed:15799966).
TRUSTED_SUSTAINABILITY

Specifications

Antigen COMMD7
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene COMMD7
Gene Accession No. Q86VX2
Gene Alias 2310010I22Rik; AW060956; C20orf92; COMM domain containing 7; COMM domain-containing protein 7; COMMD7; dJ1085F17.3; mU3; RP11-503P21.3
Gene Symbols COMMD7
Host Species Rabbit
Immunogen Recombinant Human COMMD7. Recombinant protein control fragment (Product #RP-104986).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 149951
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.