Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COMMD8 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300353AF532
Description
COMMD8 Polyclonal specifically detects COMMD8 in Human samples. It is validated for ELISA.Specifications
COMMD8 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
54951 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
Alexa Fluor 532 | |
COMM domain containing 8, COMM domain-containing protein 8, FLJ20502 | |
Produced in rabbits immunized with purified, recombinant COMMD8 (AAH19826.1; Met1-Lys183). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction