Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COMMD8 Antibody, Novus Biologicals™
SDP

Catalog No. NBP1796620 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP1796620 20 μL
NBP179661 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP1796620 Supplier Novus Biologicals Supplier No. NBP17966120UL

Rabbit Polyclonal Antibody

COMMD8 Polyclonal specifically detects COMMD8 in Human samples. It is validated for Western Blot.

Specifications

Antigen COMMD8
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. NP_060315
Gene Alias COMM domain containing 8, COMM domain-containing protein 8, FLJ20502
Gene Symbols COMMD8
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human COMMD8The immunogen for this antibody is COMMD8. Peptide sequence IAALRMPLLSLHLDVKENGEVKPYSIEMSREELQNLIQSLEAANKVVLQL.
Molecular Weight of Antigen 21 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54951
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.