Learn More
Invitrogen™ COMMD8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114095
Description
Immunogen sequence: IAALRMPLLS LHLDVKENGE VKPYSIEMSR EELQNLIQSL EAANKVVLQL For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 79%; Dog: 86%; Guinea Pig: 92%; Horse: 92%; Human: 100%; Mouse: 79%; Rabbit: 86%; Rat: 86%; Yeast: 83%.
The protein encoded by this gene binds coiled-coil domain-containing protein 22 (CCDC22), and this complex can regulate the turnover of I-kappa-B and the activation of NF-kappa-B.
Specifications
| COMMD8 | |
| Polyclonal | |
| Unconjugated | |
| COMMD8 | |
| C77770; COMM domain containing 8; COMM domain-containing protein 8; Commd8; D5Buc26e; MDS022 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 54951 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NX08 | |
| COMMD8 | |
| Synthetic peptide directed towards the middle region of human COMMD8. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.